Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral MIP-III Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $503.00
Specifications
Antigen | MIP-III |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
MIP-III Polyclonal specifically detects MIP-III in Viral samples. It is validated for Western Blot.Specifications
MIP-III | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Virus | |
E. coli-derived recombinant human herpes virus-8 MIP-III Ser27-Pro114 Accession # P88968 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects viral MIP-III in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with _______________ is observed. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P88968 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title