Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Tick-borne Encephalitis Virus NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
| Antigen | Tick-borne Encephalitis Virus Envelope |
|---|---|
| Clone | 1004134 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10106SP
![]() |
R&D Systems
MAB10106SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10106100
![]() |
R&D Systems
MAB10106100 |
100 μg |
Each for $432.50
|
|
|||||
Description
Tick-borne Encephalitis Virus NS1 Monoclonal specifically detects Tick-borne Encephalitis Virus NS1 in Viral samples. It is validated for ELISA.Specifications
| Tick-borne Encephalitis Virus Envelope | |
| ELISA | |
| Monoclonal | |
| Lyophilized | |
| Virus | |
| Human embryonic kidney cell line HEK293-derived Tick-borne Encephalitis Virus NS1, Amino Acids 777-1128 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Tick-borne Encephalitis Virus NS1 in direct ELISAs. |
| 1004134 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title