Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viral wnvNS3 Protease Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB29071
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB29071 100 μg
MAB29071SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB29071 Supplier R&D Systems Supplier No. MAB29071
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

wnvNS3 Protease Monoclonal specifically detects wnvNS3 Protease in Viral samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.

Specifications

Antigen wnvNS3 Protease NS2b cofactor
Applications Western Blot, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 345712
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. ABD85079
Host Species Mouse
Immunogen E. coli-derived recombinant wnvNS3 Protease protein Ser1423-Lys1470, Gly1506-Leu1689 Accession # ABD85079
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 912267
Test Specificity Detects West Nile Virus NS2b cofactor of the NS3 Protease complex in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile Tris-buffered saline, pH 7.3.
Target Species Virus
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.