Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Yellow Fever Virus NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB97761100
This item is not returnable.
View return policy
Description
Yellow Fever Virus NS1 Monoclonal antibody specifically detects Yellow Fever Virus NS1 in Virus samples. It is validated for ELISASpecifications
Yellow Fever Virus NS1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2b |
ELISA | |
994717 | |
ELISA | |
P03314 | |
Human embryonic kidney cell HEK293-derived Yellow Fever Virus NS1, Gln780-His1134, Accession # P03314 | |
100 μg | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction