Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viral Zika Virus (H/PF/2013) Envelope Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB100091SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB100091SP 25 μg
MB100091100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB100091SP Supplier R&D Systems Supplier No. MAB100091SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Zika Virus (H/PF/2013) Envelope Monoclonal specifically detects Zika Virus (H/PF/2013) Envelope in Viral samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Zika Virus (H/PF/2013) Envelope
Applications Western Blot, ELISA
Classification Monoclonal
Clone 990844
Conjugate Unconjugated
Dilution Western Blot 1 μg;/mL, ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Host Species Mouse
Immunogen E. coli-derived recombinant Zika Virus (H/PF/2013) Envelope, Leu590-Ser695
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Test Specificity Detects Zika Virus (H/PF/2013) Envelope (E3) in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Virus
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.