Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Zika Virus (H/PF/2013) NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB99202100
This item is not returnable.
View return policy
Description
Zika virus (H/PF/2013) NS1 Monoclonal specifically detects Zika virus (H/PF/2013) NS1 in Viral samples. It is validated for Western Blot.Specifications
Zika virus (H/PF/2013) NS1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Mouse | |
100 μg | |
Detects Viral Zika Virus (H/PF/2013) in direct ELISAs. | |
Virus | |
IgG2a |
Western Blot | |
990910 | |
Western Blot 1 μ/mL | |
AHZ13508 | |
Human embryonic kidney cell, HEK293-derived recombinant Viral Zika Virus (H/PF/2013) NS1, Val796 & Ser799-Ser1148, Accession No. AHZ13508 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction