Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Zika Virus (H/PF/2013) NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $456.00
Specifications
Antigen | Zika virus (H/PF/2013) NS1 |
---|---|
Clone | 990907 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB99201SP
![]() |
R&D Systems
MAB99201SP |
25 μg |
Each for $200.50
|
|
|||||
MAB99201100
![]() |
R&D Systems
MAB99201100 |
100 μg |
Each for $456.00
|
|
|||||
Description
Zika virus (H/PF/2013) NS1 Monoclonal specifically detects Zika virus (H/PF/2013) NS1 in Viral samples. It is validated for ELISA.Specifications
Zika virus (H/PF/2013) NS1 | |
ELISA | |
Monoclonal | |
Supernatant | |
Virus | |
AHZ13508 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
990907 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Human embryonic kidney cell, HEK293-derived recombinant Viral Zika Virus (H/PF/2013) NS1, Val796 & Ser799-Ser1148, with a C-terminal 6-His tag, Accession No. AHZ13508 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects Viral Zika Virus (H/PF/2013) NS1 in direct ELISAs |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title