Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Virex™ II 256 One-Step Disinfectant Cleaner and Deodorant

Catalog No. p-7221808
Click to view available options
Volume (Metric):
946 mL
2.5 L
3.78 L
This item is not returnable. View return policy
This item is not returnable. View return policy

A one-step, quaternary disinfectant cleaner and deodorant to clean and disinfect hard surfaces.

  • Kills microorganisms including HIV-1, VRE, MRSA, GRSA, MRSE, VISA, PRSP, Herpes Simplex Types 1&2, Influenza Type A2, Adenovirus, Rotavirus and many more
  • Kills the organisms that cause odors and works as an odor counteractant to further eliminate odors not associated with bacteria
  • Highly concentrated quaternary formula provides excellent one-step, cost-effective cleaning and disinfection

Specifications

Product Type Quat-based Disinfectant
pH 10.2
Sterility Non-sterile
Certifications/Compliance EPA, FDA
Container Type Jug
For Use With (Application) Medical and general institutional facilities general purpose
Quantity 4 Pk.
Volume (English) 1 gal.
Volume (Metric) 3.78 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.