Learn More
Description
Used for: endosomal uptake assays, protein delivery experiments and evaluation of protein cargo internalization.
Application: Live-cell imaging as a self-labeling tag substrate; fluorescent microscopy and flow cytometry.
Properties and Photophysical Data: shows a high degree of fluorogenicity. Excitation and emission maxima (λ) are 555 nm and 581 nm, respectively; quantum yield = 0.40; extinction coefficient = 90,900 M-1cm-1; pH sensitive fluorescence h = 1.46. PKa = 6.25. Colourless and non-fluorescent at low pH.
Specifications
Specifications
| Chemical Name or Material | 4-((2-(2-((6-Chlorohexyl)oxy)ethoxy)ethyl)carbamoyl)-2-(2,7-difluoro-6-hydroxy-10,10-dimethyl-3-oxo-3,10-dihydroanthracen-9-yl)benzoic acid |
| Quantity | 10 μg |
| Product Line | Tocris |
| Target | Self-labeling Tags |
| Molecular Formula | C34H36ClF2NO7 |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
