Learn More
Virginiamycin Polyclonal Antibody

Description
Virginiamycin Polyclonal Antibody for ELISA
Specifications
Specifications
| Antigen | Virginiamycin |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.3 |
| Host Species | Sheep |
| Immunogen | Virginiamycin M-BSA. |
| Purification Method | Purified |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.