Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Virilizer Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP121363R
Description
Virilizer Polyclonal antibody specifically detects Virilizer in Human samples. It is validated for Western BlotSpecifications
Virilizer | |
Polyclonal | |
Western Blot | |
DKFZP434I116, DKFZp781B2117, fSAP121, functional spliceosome-associated protein 121, KIAA1429, MGC138493, MGC141940, protein virilizer homolog | |
The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human Virilizer (KIAA1429) using the numbering given in entry NP_056311.2 (GeneID 25962). | |
0.1 mL | |
Cell Biology | |
25962 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction