Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
visinin-like 1 Antibody (3A9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225167SS
Description
visinin-like 1 Monoclonal specifically detects visinin-like 1 in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
visinin-like 1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000 | |
P62760 | |
VSNL1 | |
Full length human visinin-like 1 [UniProt# P62760] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Bovine, Mouse and Rat. Other species have not been tested. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
3A9 | |
Unconjugated | |
50% PBS, 50% glycerol with 5mM Sodium Azide | |
HLP3Hippocalcin-like protein 3, HPCAL3, HUVISL1, VILIP-1, VILIPVLP-1, visinin-like 1, visinin-like protein 1, VISL1 | |
Mouse | |
22 kDa | |
0.025 mL | |
Stem Cell Markers | |
7447 | |
Human, Mouse, Rat, Pig, Bovine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction