Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
visinin-like 1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300332B
Description
visinin-like 1 Polyclonal specifically detects visinin-like 1 in Human samples. It is validated for ELISA.Specifications
| visinin-like 1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Biotin | |
| HLP3Hippocalcin-like protein 3, HPCAL3, HUVISL1, VILIP-1, VILIPVLP-1, visinin-like 1, visinin-like protein 1, VISL1 | |
| Produced in rabbits immunized with purified, recombinant visinin-like 1 (P62760; Gly2-Lys191). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 7447 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction