Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
visinin-like 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | visinin-like 1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:10000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP225169SS
![]() |
Novus Biologicals
NBP225169SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP225169
![]() |
Novus Biologicals
NBP225169 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
visinin-like 1 Polyclonal specifically detects visinin-like 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.Specifications
visinin-like 1 | |
Western Blot 1:10000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000 | |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
50% PBS, 50% Glycerol with 5mM Sodium Azide | |
HLP3Hippocalcin-like protein 3, HPCAL3, HUVISL1, VILIP-1, VILIPVLP-1, visinin-like 1, visinin-like protein 1, VISL1 | |
VSNL1 | |
IgG | |
Affinity Purified | |
Human, Bovine, Mouse and Rat. Other species have not been tested. |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
P62760 | |
7447 | |
Full length recombinant human VLP1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
22 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title