Learn More
MilliporeSigma™ Supelco™ Visiprep™ SPE Vacuum Manifold, Standard
Catalog No. 111020696 Shop All MilliporeSigma Supelco Products
Click to view available options
No. of Ports:
12
24
Catalog No. 11-102-0696
Supplier: MilliporeSigma™ Supelco™ 57250U
Only null left
Add to Cart
Add to Cart
Unlike other vacuum manifolds, the Visiprep system contains a patented valve system that allows for precise flow control through each SPE tube via rotating, independent, screw-type valves situated in each port within the manifold cover.
SPE is a form of chromatography, and, as with other chromatographic techniques, control of flow rate is critical for maintaining reproducible extractions. Unlike other vacuum manifolds, the Visiprep system contains a patented valve system that allows for precise flow control through each SPE tube via rotating, independent, screw-type valves situated in each port within the manifold cover. Visiprep vacuum manifolds allow you to process up to 12 (12-port version) or 24 (24-port version) SPE samples simultaneously. Spacing between ports are slightly smaller on the 24-port versions than the 12-port versions so consideration of tube sizes is recommended.
Features and Benefits:
  • Patented screw-type valves within each SPE port for precise flow control
  • Glass basin will not dissolve, fog, or discolor when exposed to solvents
  • Legs on cover allow user to easily rest cover on work surface when removed from the manifold
  • Screw-type solvent-resistant vacuum bleed gauge and valve offer better sealing and vacuum control. Valve takes 1/4 in.
  • vacuum tubing.
  • PP collection vessel rack accommodates autosampler vials; small scintillation vials (22.75 mm O.D. recommended); 10 and 16 mm test tubes; and 1, 2, 5, and 10 mL volumetric flasks. An optional plate for 20 mL scintillation vials is available for 12-port models.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Food and Beverages
Product Line Visiprep
Type SPE Manifold
No. of Ports 24
Visiprep is a trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.