Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Vismodegib
GREENER_CHOICE

Catalog No. 771010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-101-0 10 mg
77-105-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 77-101-0 Supplier Tocris Bioscience™ Supplier No. 7710/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent hedgehog pathway inhibitor

Vismodegib is a potent hedgehog pathway inhibitor (IC50 = 3 nM). It causes tumor regressions in allograft medulloblastoma mice model and tumor growth inhibition in a xenograft mouse models of colorectal cancer. Vismodegib is orally bioavailable.

Specifications

Chemical Name or Material 2-Chloro-N-[4-chloro-3-(2-pyridinyl)phenyl]-4-(methylsulfonyl)benzamide
CAS 879085-55-9
Quantity 10 mg
Target Hedgehog Signaling Inhibitors
Molecular Formula C19H14Cl2N2O3S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.