Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VISTA/B7-H5/PD-1H Antibody (1011484), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB71267PEATT594
Description
VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Flow CytometrySpecifications
VISTA/B7-H5/PD-1H | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1011484 | |
Flow Cytometry | |
B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
Mouse myeloma cell line NS0-derived recombinant human VISTA/B7-H5/PD-1H, Phe33-Ala194, Accession # Q9H7M9 | |
0.1 mL | |
Cancer, Cardiovascular Biology, Immunology | |
64115 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction