Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VISTA/B7-H5/PD-1H Antibody (730801), DyLight 680, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB71262FR
Description
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Western Blot.Specifications
VISTA/B7-H5/PD-1H | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C10ORF54 | |
Partial recombinant human VISTA/B7-H5 (amino acids 33-194) [UniProt Q9H7M9] | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot | |
730801 | |
Western Blot | |
B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
Mouse | |
Protein A or G purified | |
RUO | |
64115 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction