Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VISTA/B7-H5/PD-1H Antibody (855328), DyLight 405, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB9057V
Description
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
VISTA/B7-H5/PD-1H | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C10ORF54 | |
Partial recombinant human VISTA/B7-H5 protein (aa 33-194) | |
100 μL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
855328 | |
Flow Cytometry, Flow (Cell Surface) | |
B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
Mouse | |
Protein A or G purified | |
RUO | |
64115 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction