Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VISTA/B7-H5/PD-1H Antibody (MH5A), DyLight 594, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP311969DL594
Description
VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Mouse samples. It is validated for ELISA, ImmunohistochemistrySpecifications
VISTA/B7-H5/PD-1H | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Armenian Hamster | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
IgG λ |
ELISA, Immunohistochemistry | |
MH5A | |
ELISA, Immunohistochemistry | |
B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
The antibody was generated by immunising Armenian hamsters with mVISTA/B7-H5/PD-1HIg. | |
0.1 mL | |
Cancer, Cardiovascular Biology, Immunology | |
64115 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction