Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VISTA/B7-H5/PD-1H Antibody (MH5A), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP311973MFV450
Description
VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, NeutralizationSpecifications
| VISTA/B7-H5/PD-1H | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Neutralization | |
| MH5A | |
| ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Block/Neutralize | |
| B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| The antibody was generated by immunising Armenian hamsters with mVISTA/B7-H5/PD-1HIg. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Immunology | |
| 64115 | |
| Store at 4C in the dark. | |
| IgG λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction