Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VISTA/B7-H5/PD-1H Rabbit anti-Mouse, Clone: mam82, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | VISTA/B7-H5/PD-1H |
---|---|
Clone | mam82 |
Concentration | 1 mg/mL |
Dilution | ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Block/Neutralize |
Applications | ELISA, Immunohistochemistry, Immunofluorescence, Neutralization |
Description
VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, NeutralizationSpecifications
VISTA/B7-H5/PD-1H | |
1 mg/mL | |
ELISA, Immunohistochemistry, Immunofluorescence, Neutralization | |
Unconjugated | |
RUO | |
Mouse | |
B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
The antibody was generated by immunising a VISTA/B7-H5/PD-1H-KO mouse with mouse VISTA/B7-H5/PD-1H Ig fusion protein and adjuvant. | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
mam82 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Block/Neutralize | |
Monoclonal | |
Rabbit | |
Cancer, Cardiovascular Biology, Immunology | |
PBS | |
64115 | |
IgG κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title