Learn More
Invitrogen™ VISTA Chimeric Recombinant Rabbit Monoclonal Antibody (MH5A)

Description
Specificity: This antibody specifically binds PD-1H.
Specifications
Specifications
| Antigen | VISTA Chimeric |
| Applications | ELISA, Functional Assay, Inhibition Assays, Immunohistochemistry, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | MH5A |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | VSIR |
| Gene Accession No. | Q9D659 |
| Gene Alias | 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.