Learn More
Invitrogen™ VISTA Monoclonal Antibody (B7H5DS8), PE, eBioscience™, Invitrogen™
Description
Description: This B7H5DS8 monoclonal antibody recognizes human VISTA, also known as B7-H5 or PD-1H. This clone has been tested and shown not to cross-react with human B7-H4, B7-H6 and B7-H7. This B7H5DS8 antibody will work in flow cytometry only on live and not paraformaldehyde-fixed cells. Applications Reported: This B7H5DS8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B7H5DS8 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
Specifications
Specifications
| Antigen | VISTA |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | B7H5DS8 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | VSIR |
| Gene Accession No. | Q9H7M9 |
| Gene Alias | 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.