Learn More
Invitrogen™ VISTA Monoclonal Antibody (MH5A), Functional Grade, eBioscience™, Invitrogen™

Description
Description: The monoclonal antibody MH5A reacts with a homolog of PD-1 (PD-1H) also known as VISTA. PD-1H is a 309 aa type I transmembrane protein that belongs to the CD28 superfamily. This cell surface receptor is expressed on nearly all hematopoietic cells except B cells. VISTA negatively regulates T cell responses by suppressing early TCR activation, inhibiting cell proliferation, and suppressing TH1 cytokine production. The MH5A antibody has been reported to inhibit T cell infiltration in graft-versus-host disease mouse models. Applications Reported: This MH5A antibody has been reported for use in flow cytometric analysis. Applications Tested: This MH5A antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
Specifications
Specifications
| Antigen | VISTA |
| Applications | Control, Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Clone | MH5A |
| Concentration | 1 mg/mL |
| Conjugate | Functional Grade |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | VSIR |
| Gene Accession No. | Q9D659 |
| Gene Alias | 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.