Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA Mouse anti-Human , Alexa Fluor 647, Clone: MIH65.rMAb (also known as MIH65), BD Biosciences™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 566671

Catalog No. BDB566671


Only null left
Add to Cart

Description

Description

VISTA Monoclonal antibody specifically detects VISTA in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VISTA
Monoclonal
Alexa Fluor 647
VSIR; B7-H5; B7H5; C10orf54; DD1alpha; GI24; SISP1
Human VISTA Transfected Cell Line
RUO
Human
IgG1 κ
Flow Cytometry
MIH65.rMAb (also known as MIH65)
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.