Learn More
VISTA Mouse anti-Human , PE, Clone: MIH65.rMAb (also known as MIH65), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | VISTA |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MIH65.rMAb (also known as MIH65) |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | VSIR; B7-H5; B7H5; C10orf54; DD1alpha; GI24; SISP1 |
| Host Species | Mouse |
| Immunogen | Human VISTA Transfected Cell Line |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.