Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA Rat anti-Mouse, Brilliant Violet 421, Clone: MIH64, BD

Catalog No. BDB566271 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566271 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566271 Supplier BD Biosciences Supplier No. 566271
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MIH64 monoclonal antibody specifically binds to V-domain Ig suppressor of T cell activation (VISTA) which is also known as, Differentiation of ESCs 1 (Dies1). VISTA is a type I transmembrane glycoprotein that belongs to the Ig superfamily. VISTA has homology to the B7 family ligand, PD-L1 (CD274), and is likewise known as PD-1H. VISTA is expressed on naοve and activated T cells, natural killer (NK) cells, macrophages, dendritic cells, and neutrophils. VISTA functions as an inhibitory ligand. It can serve as a negative immune-checkpoint protein that suppresses T cell cytokine production and proliferation. It is also involved in regulating stem cell differentiation.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VISTA
Classification Monoclonal
Clone MIH64
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Vista, PD-1H, Dies1
Host Species Rat
Immunogen Mouse
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.