Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA Rat anti-Mouse, Brilliant Violet 786, Clone: MIH64, BD Optibuild™

Catalog No. BDB742725 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742725 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742725 Supplier BD Biosciences Supplier No. 742725
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MIH64 monoclonal antibody specifically binds to V-domain Ig suppressor of T cell activation (VISTA) which is also known as, Differentiation of ESCs 1 (Dies1). VISTA is a type I transmembrane glycoprotein that belongs to the Ig superfamily. VISTA has homology to the B7 family ligand, PD-L1 (CD274), and is likewise known as PD-1H. VISTA is expressed on naοve and activated T cells, natural killer (NK) cells, macrophages, dendritic cells, and neutrophils. VISTA functions as an inhibitory ligand. It can serve as a negative immune-checkpoint protein that suppresses T cell cytokine production and proliferation. It is also involved in regulating stem cell differentiation.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VISTA
Classification Monoclonal
Clone MIH64
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Vista, PD-1H, Dies1
Host Species Rat
Immunogen Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.