Learn More
VistaLab™ Technologies MLA™ Brand Precision Pipetters


Shop All VistaLab Technologies Inc. Products



Description
- 'To deliver' capability ensures volume accuracy over a wide range of viscosities
- Ideal for repetitive delivery of reagents when high precision and accuracy are required
- Sleeves are color coded for easy volume identification
- Capacity engraved on bonnet
Order Info
Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Accuracy | ±1.00% |
| Autoclavable | Not Autoclavable |
| Color | Brown |
| Disposable | Reusable |
| For Use With | Micropipette Tip |
| Material | Aluminum, Stainless Steel |
| Model | F1 |
| Number of Channels | 1 |
| Sterility | Non-sterile |
| Type | Precision Pipette |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.