Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VistaLab™ Technologies Reagent Reservoirs: 12-Channel

Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
Bulk
Individually Wrapped
Quantity:
10/Cs.
25/Cs.
Description
- Polystyrene with trimmed base corners
- Distinct pour-off spouts to empty reservoirs without spills
- Well-marked graduated increments on inside wall minimize reagent waste
- Wide base and sturdy construction provide stability and help to avoid spills
- Molded corner structures allow easy separation of bulk packaged reservoirs
- 100mL model (Cat. No. 21-382-111) supplied with lid
- Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
- 5 mL capacity / well
Specifications
Specifications
Certifications/Compliance | RNase/DNase, Pyrogen-free |
Color | Clear |
Capacity (Metric) | 60 mL |
Material | Polypropylene |
Sterility | Sterile |
Type | Reagent Reservoir |
For Use With (Application) | General Liquid Handling |
Height (English) | 4 in. |
Includes | Lid |
Length (English) | 11 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction