Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Reagent Reservoirs
GREENER_CHOICESDP

Catalog No. 21385100
Click to view available options
Capacity (Metric):
10 mL
100 mL
100 mL (with lid)
1800 mL
25 mL
25 mL (divided)
50 mL
Sterility:
Non-sterile
Sterile
Packaging:
Bulk
Individually Wrapped
Individually packed
Quantity:
10/bag x 10; 100/Cs.
100/Cs.
20/bag x 10; 200/Cs.
200 in. 20 bags of 10
50/Cs.
Each

Unique trough-within-trough design maximizes reagent recovery

  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
  • 100mL model (Cat. No. 21-385-111) is supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile

Order Info

Reservoir lid sold separately (Cat. No. 21-385-110).

Specifications

Certifications/Compliance Pyrogen-free
Color White
Capacity (Metric) 100 mL
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) General Liquid Handling
Height (English) 6 in.
Length (English) 18 in.
Packaging Bulk
Quantity 20/bag x 10; 200/Cs.
Width (English) 12 in.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.