Learn More
VistaLab™ Technologies Seal Kits for MLA™ Brand Pipets
SDP
Catalog No. p-3237905 Shop All VistaLab Technologies Inc. Products
Click to view available options
Volume (Metric):
121 to 200 μL
201 to 350 μL
31 to 120 μL
351 to 1000 μL
5 to 30 μL
Provide materials for maintenance of MLA D-Tipper™, Precision, and two-stroke pipetters
Each kit contains seals and instructions
Order Info
Lubricant sold separately (P/N 22-146-318)
Specifications
For Use With Pipettes
Material Plastic
Volume (Metric) 5 to 30 μL
For Use With (Equipment) 5 to 30 μL MLA™ D-Tipper™, Precision and Two Stroke Pipette
Includes Lubricant, seals and instructions
Quantity 1 Ea.
Length (English) 2 in.
Width (English) 1 in.
Height (English) 1 in.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.