Learn More
Eisco™ Visual Scientifics Car Kit
SCI_ED
Catalog No. s23761
Click to view available options
:
Visual Scientifics Car Kit
For investigating static and kinetic friction, dynamic equilibrium, unbalanced forces and work-energy theory.
  • For use with PTIPCR incline plane and car
  • Car can be used with plane to predict velocity and quantify friction for further study
  • Includes instructions for classroom and lab use
Specifications
Product Type Kit, Visual Scientifics Car
For Use With (Application) For investigating static and kinetic friction, dynamic equilibrium, unbalanced forces and work-energy theory
For Use With (Equipment) Plane to predict velocity and quantify friction for further study
Includes PTIPCR incline plane and car, instructions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.