Learn More
Description
Includes
Substrate A, 100mL; Substrate B, 100mL
- High sensitivity – femtogram level detection
- High signal intensity
- Excellent signal to noise ratio
- Compatible with X-ray film or CCD imaging systems
- Store unopened product at 2° to 8°C. Do not use past expiration date.
Specifications
Specifications
| Description | General Laboratory Research Reagent |
| Product Type | ECL Western Blot Substrate |
| Quantity | 200 mL (2 x 100 mL) |
| Content And Storage | Substrate A (100mL), Substrate B (100mL). Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
