Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin A Acetate, USP, Spectrum™ Chemical
SDP

Catalog No. 18607709
Click to view available options
Quantity:
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle

V1257, 127-47-9, C22H32O2

Spectrum™ Chemical Vitamin A Acetate, USP is also known as retinyl acetate or retinol acetate and is often used to fortify foods with vitamin A. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-70-8, 9005-25-8, 127-47-9, 57-50-1, 128-37-0
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.50
InChI Key QGNJRVVDBSJHIZ-QHLGVNSISA-N
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate
SMILES CC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C

Specifications

CAS Max % 40 to 60%, 20 to 40%, 10 to 20%, 10 to 20%, 1 to <3%
Packaging Amber Glass Bottle
Quantity 500 g
Grade USP
Physical Form Crystals or Flakes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.