Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin A Acetate, USP, Spectrum™ Chemical
SDP

Catalog No. 18607710 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-710 25 kg Amber Glass Bottle
18-607-708 100 g Amber Glass Bottle
18-607-709 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-710 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. V125725KGBL

missing translation for 'validMainframeMsg'

V1257, 127-47-9, C22H32O2

Spectrum™ Chemical Vitamin A Acetate, USP is also known as retinyl acetate or retinol acetate and is often used to fortify foods with vitamin A. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-70-8 , 9005-25-8 , 127-47-9 , 57-50-1 , 128-37-0
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.50
InChI Key QGNJRVVDBSJHIZ-QHLGVNSISA-N
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate
SMILES CC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C

Specifications

CAS Max % 40 to 60%, 20 to 40%, 10 to 20%, 10 to 20%, 1 to <3%
Packaging Amber Glass Bottle
Quantity 25 kg
Grade USP
Physical Form Crystals or Flakes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.