Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vitamin B1 Monoclonal Antibody (1C6)
GREENER_CHOICE

Catalog No. PIMA549651
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549651 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549651 Supplier Invitrogen™ Supplier No. MA549651
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Vitamin B1, also known as thiamine, is a water-soluble vitamin that is essential for the proper functioning of the body. It is involved in a variety of metabolic processes, including the production of energy from food, the synthesis of neurotransmitters, and the maintenance of a healthy nervous system. Vitamin B1 is found in a variety of foods, including whole grains, meat, fish, legumes, and nuts. Vitamin B1 deficiency can lead to a range of symptoms, including fatigue, muscle weakness, and nerve damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vitamin B1
Applications ELISA, Immunohistochemistry, Lateral Flow
Classification Monoclonal
Clone 1C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias thiamin; thiamine
Host Species Mouse
Immunogen Conjugate of VB1 hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.