Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vitamin B12 Monoclonal Antibody (2F9)

Catalog No. PIMA549624
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549624 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549624 Supplier Invitrogen™ Supplier No. MA549624

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Vitamin B12, also known as cobalamin, is a water-soluble vitamin that plays a crucial role in the proper functioning of the brain, nervous system, and the formation of red blood cells. It is primarily found in animal products such as meat, fish, and dairy, and is essential for the maintenance of good health. Vitamin B12 is necessary for the conversion of food into energy and aids in the synthesis of DNA and RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vitamin B12
Applications ELISA, Lateral Flow, Lateral Flow
Classification Monoclonal
Clone 2F9
Concentration 6.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias cyanocobalamin
Host Species Mouse
Immunogen Conjugate of VB12 hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.