Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vitamin B12a hydrochloride, ≥95%, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
100 mg
1 g
Description
Vitamins, Nutraceuticals and Natural Products

Specifications
Specifications
Packaging | Glass Bottle |
CAS | 59461-30-2 |
Physical Form | Neat |
Linear Formula | C62H89 CoN13O15 P · HCl |
MDL Number | MFCD00167463 |
Synonym | Hydroxocobalamin hydrochloride |
Type | Analytical Standard |
Quantity | 100 mg |
Formula Weight | 1382.82 |
Percent Purity | ≥95% |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction