Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vitamin B2 Monoclonal Antibody (3H8)
GREENER_CHOICE

Catalog No. PIMA549650
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549650 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549650 Supplier Invitrogen™ Supplier No. MA549650
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Vitamin B2, also known as riboflavin, is a water-soluble vitamin that is essential for the proper functioning of the body. It is involved in a variety of metabolic processes, including the production of energy from food, the synthesis of red blood cells, and the maintenance of healthy skin, eyes, and nervous system. Vitamin B2 is found in a variety of foods, including dairy products, meat, fish, whole grains, and leafy green vegetables. Vitamin B2 deficiency is rare, but it can lead to a range of symptoms, including anemia, skin disorders, and eye problems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vitamin B2
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Riboflavin
Host Species Mouse
Immunogen Conjugate of VB2 hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.