Learn More
Invitrogen™ Vitamin B5 Monoclonal Antibody (2A2)

Description
Antibody tested in indirect ELISA.
Specifications
Specifications
| Antigen | Vitamin B5 |
| Applications | ELISA, Immunohistochemistry, Lateral Flow, SDS-Page |
| Classification | Monoclonal |
| Clone | 2A2 |
| Concentration | 3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene Alias | Pantothenic Acid |
| Host Species | Mouse |
| Immunogen | VB5-hapten-EDC-BSA. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.