Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin D binding protein Mouse anti-Human, Clone: 1E4D10, Proteintech
SDP

Catalog No. 501736609 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6609 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6609 Supplier Proteintech Group Inc Supplier No. 661751IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.

Specifications

Antigen Vitamin D binding protein
Applications Western Blot
Classification Monoclonal
Clone 1E4D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GC
Gene Accession No. P02774
Gene Alias DBP, DBP/GC, GC, Gc globulin, VDBP,GC, Vitamin D binding protein
Gene Symbols GC
Host Species Mouse
Immunogen Vitamin D binding protein Fusion Protein Ag9803
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2638
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.