Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vitamin D BP Mouse anti-Human, Clone: VDBP/4482, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP31392420UG
This item is not returnable.
View return policy
Description
Vitamin D BP Monoclonal antibody specifically detects Vitamin D BP in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
Vitamin D BP | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P02774 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Peptide Array | |
VDBP/4482 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
DBP/GC, DBPVDBG, EC 6.3.1.5, Gc-globulin, GRD3, Group-specific component, group-specific component (vitamin D binding protein), hDBP, VDB, VDBP, vitamin D-binding alpha-globulin, vitamin D-binding protein | |
Recombinant fragment (around aa 35-175) of human Vitamin D BP protein (exact sequence is proprietary) (Uniprot: P02774) | |
20 μg | |
Cancer | |
2638 | |
Store at 4°C. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction