Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Vitamin D Free Diet, Powder

Vitamin D free diet. Contains vitamins and minerals.

$713.00 - $2706.00

Specifications

Product Type Animal Diet
Autoclavable Not Autoclavable
Disposable Single-use
Format Powder
Sterility Non-sterile
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP296007310
View Documents
MP Biomedicals
0296007310
10 kg
Each for $713.00
Only null left
Add to Cart
 
MP296007320
View Documents
MP Biomedicals
0296007320
20 kg
Each for $1,248.00
Only null left
Add to Cart
 
MP296007350
View Documents
MP Biomedicals
0296007350
50 kg
Each for $2,706.00
Only null left
Add to Cart
 
Description

Description

Diet consisting of vitamin free casein, whole wheat flour, corn oil, alphacel (non-nutritive), calcium carbonate and calcium phosphorous free salt mixture. The diet contains all nutrients (except vitamin D) required by the rat, and when cholecalciferol is added (5 IU/gm diet), the diet provides growth and reproduction equal to that of commercial stock diets. Using whole wheat as the source of carbohydrate the rats experience better growth and development than with mono or disaccharides. There is some evidence, however, to suggest that vitamin D depletion may occur more rapidly with sucrose as the carbohydrate source. The calcium content of the diet can be varied by changing the amount of CaCO3 added.

Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Not Autoclavable
Powder
Refrigerated
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.