Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, 95%
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 54.0°C to 56.0°C |
| Assay Percent Range | 18.46 to 20.82% (ICP on dried sample) |
| Packaging | Glass bottle |
| Quantity | 50 g |
| Solubility Information | Solubility in water: soluble. |
| Formula Weight | 468.28 |
| Percent Purity | 95% |
| Chemical Name or Material | Ethylene glycol-bis(β-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt |
Sécurité et manipulation
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'
missing translation for 'spotOppurtunityImprovement'missing translation for 'shareContentCorrection'