Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vitamin D3 Antibody (685503), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6566P
Description
Vitamin D3 Monoclonal antibody specifically detects Vitamin D3 in Broad Species Reactivity samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Vitamin D3 | |
Monoclonal | |
PE | |
PBS | |
25-Hydroxyvitamin D3 | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry (Intracellular Staining), CyTOF | |
685503 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Broad Species Reactivity | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction