Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vitamin D3 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC6566V100UG
This item is not returnable.
View return policy
Description
Vitamin D3 Monoclonal specifically detects Vitamin D3 in Multi-Species samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Vitamin D3 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Mouse | |
100 μg | |
Primary | |
Multi-species | |
IgG1 |
Flow Cytometry | |
685503 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
25-Hydroxyvitamin D 3 | |
RUO | |
Detects Vitamin D3 in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction