Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin E acetate, 97%

Catalog No. p-3346685
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 7695-91-2 | C31H52O3 | 472.754 g/mol

Vitamin E acetate is used as a fortifying agent and dietary supplement in food. It is used in dermatological products such as skin creams. It is an alternative to tocopherol and provides protection against the sun’s ultraviolet rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin E acetate is used as a fortifying agent and dietary supplement in food. It is used in dermatological products such as skin creams. It is an alternative to tocopherol and provides protection against the sun’s ultraviolet rays.

Solubility
Immiscible with water.

Notes
Store in a cool place. Air and light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7695-91-2
Molecular Formula C31H52O3
Molecular Weight (g/mol) 472.754
MDL Number MFCD00072042
InChI Key ZAKOWWREFLAJOT-CEFNRUSXSA-N
Synonym vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ
PubChem CID 86472
ChEBI CHEBI:32321
IUPAC Name [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate
SMILES CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Melting Point °C to 28°C
Density 0.960
Boiling Point >300°C
Flash Point 210°C (410°F)
Assay Percent Range 97%
Refractive Index 1.496
Quantity 25 g
Beilstein 97512
Sensitivity Air and light sensitive
Merck Index 14,9495
Solubility Information Immiscible with water.
Formula Weight 472.76
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Vitamin E Acetate
Show More Show Less
EINECSNumber 231-710-0
RTECSNumber GA8747000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.