Vitamin E Acetate, 97%, Alfa Aesar™

Manufacturer: Alfa Aesar A1450514

Quantity 25g

 View more versions of this product

Catalog No. AAA1450514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7695-91-2
    Physical Form Liquid
    Assay Percent Range 97%
    Molecular Formula C31H52O3
    Formula Weight 472.76
    Chemical Name or Material Vitamin E Acetate

    Specifications

    Formula Weight 472.76
    Quantity 25g
    TSCA TSCA
    MDL Number MFCD00072042
    Beilstein 97512
    Merck Index 14,9495
    RTECS Number GA8747000
    Physical Form Liquid
    Density 0.960
    Molecular Formula C31H52O3
    Assay Percent Range 97%
    CAS 7695-91-2
    Chemical Name or Material Vitamin E Acetate
    Assay 97%

    Description

    Applications
    Vitamin E acetate is used as a fortifying agent and dietary supplement in food. It is used in dermatological products such as skin creams. It is an alternative to tocopherol and provides protection against the sun’s ultraviolet rays.

    Solubility
    Immiscible with water.

    Notes
    Store in a cool place. Air and light sensitive. Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit